Assay Details
| Gene Symbol: GUCY2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000180.3 | NP_000171.1 | 11 - 12 | 2338 | 73 | |
| XM_011523816.1 | XP_011522118.1 | 10 - 11 | 2264 | 73 | ||
| GenBank mRNA | M92432.1 | - | 11 - 12 | 2337 | 73 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000123.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cGMP biosynthetic process
protein phosphorylation
receptor guanylyl cyclase signaling pathway
visual perception
regulation of rhodopsin mediated signaling pathway
intracellular signal transduction
adenylate cyclase activity
guanylate cyclase activity
protein kinase activity
receptor activity
protein binding
ATP binding
GTP binding
cGMP biosynthetic process
protein phosphorylation
receptor guanylyl cyclase signaling pathway
visual perception
regulation of rhodopsin mediated signaling pathway
intracellular signal transduction
adenylate cyclase activity
guanylate cyclase activity
protein kinase activity
receptor activity
protein binding
ATP binding
GTP binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | ENST00000574510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | ENST00000574510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | ENST00000574510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | ENST00000574510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17001819.hg.1 | ENST00000254854.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17001819.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17001819.hg.1 | TC17000123.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | ENST00000574510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | ENST00000574510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | ENST00000574510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | ENST00000574510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700006784.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700147834.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700147834.hg.1 | NM_000180.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700147834.hg.1 | TC1700006784.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700147834.hg.1 | ENST00000254854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700147834.hg.1 | NM_000180.3 | Refseq |

GUCY2D
Genomic Map