Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005557.hg.1 |
TC14002301.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005557.hg.1 |
ENST00000267568.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005557.hg.1 |
ENST00000553813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005557.hg.1 |
ENST00000555228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005557.hg.1 |
ENST00000555661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005557.hg.1 |
NM_001146154.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005557.hg.1 |
NM_001146155.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005557.hg.1 |
NM_152444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179811.hg.1 |
ENST00000267568.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179811.hg.1 |
ENST00000553813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179811.hg.1 |
ENST00000555228.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179811.hg.1 |
ENST00000556219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179811.hg.1 |
NM_001146154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179811.hg.1 |
NM_001146155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179811.hg.1 |
NM_152444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179811.hg.1 |
ENST00000267568.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179811.hg.1 |
ENST00000553813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179811.hg.1 |
ENST00000555228.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179811.hg.1 |
ENST00000556219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179811.hg.1 |
NM_001146154.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179811.hg.1 |
NM_001146155.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179811.hg.1 |
NM_152444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179811.hg.1 |
TC1400010624.hg.1 |
Clariom D TC |