| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000373127.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000373127.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000373127.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000373127.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000373127.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000373127.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000373127.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000373127.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000373127.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000426005.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000426005.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000426005.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000426005.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000426005.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000426005.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000426005.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000426005.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000426005.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000437044.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000437044.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000437044.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000437044.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000437044.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000437044.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000437044.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000437044.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000590581.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000590581.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000590581.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000590581.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000590581.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
ENST00000590581.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_001271807.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_001271807.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_001271807.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_001271807.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_001271807.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_001271807.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_001271807.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_001271807.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_001271807.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_001271807.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_001271808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_001271808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_001271808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_001271808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_001271808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_001271808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_001271808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_001271808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_001271808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_001271808.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_178174.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_178174.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_178174.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_178174.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_178174.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_178174.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_178174.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_178174.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_178174.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_178174.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011770.hg.1 |
NM_178174.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020427.hg.1 |
ENST00000373127.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020427.hg.1 |
ENST00000426005.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020427.hg.1 |
ENST00000437044.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020427.hg.1 |
NM_178174.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020427.hg.1 |
TC06001712.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000373127.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000373127.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000373127.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000373127.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000373127.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000373127.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000373127.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000373127.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000373127.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000426005.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000426005.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000426005.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000426005.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000426005.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000426005.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000426005.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000426005.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000426005.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000437044.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000437044.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000437044.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000437044.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000437044.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000437044.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000437044.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000437044.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000590581.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000590581.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000590581.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000590581.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000590581.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
ENST00000590581.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_001271807.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_001271807.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_001271807.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_001271807.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_001271807.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_001271807.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_001271807.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_001271807.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_001271807.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_001271807.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_001271808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_001271808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_001271808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_001271808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_001271808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_001271808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_001271808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_001271808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_001271808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_001271808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_178174.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_178174.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_178174.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_178174.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_178174.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_178174.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_178174.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_178174.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_178174.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_178174.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011770.hg.1 |
NM_178174.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180826.hg.1 |
ENST00000373127.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180826.hg.1 |
ENST00000426005.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180826.hg.1 |
ENST00000437044.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180826.hg.1 |
ENST00000590581.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180826.hg.1 |
NM_001271807.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180826.hg.1 |
NM_001271808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180826.hg.1 |
NM_178174.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180826.hg.1 |
ENST00000373127.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180826.hg.1 |
ENST00000426005.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180826.hg.1 |
ENST00000437044.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180826.hg.1 |
ENST00000590581.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180826.hg.1 |
NM_001271807.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180826.hg.1 |
NM_001271808.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180826.hg.1 |
NM_178174.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180826.hg.1 |
TC0600011770.hg.1 |
Clariom D TC |