Assay Details
| Gene Symbol: OACYLP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DV080046.1 | - | 2 - 3 | 199 | 98 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800009235.hg.1 | ENST00000587689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009235.hg.1 | ENST00000587689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009235.hg.1 | ENST00000587689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009235.hg.1 | ENST00000587689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009235.hg.1 | ENST00000587689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009235.hg.1 | ENST00000587790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009235.hg.1 | ENST00000587790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009235.hg.1 | ENST00000587790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009235.hg.1 | ENST00000587790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009235.hg.1 | ENST00000587790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009235.hg.1 | ENST00000587790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009235.hg.1 | ENST00000587790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009235.hg.1 | ENST00000587790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009235.hg.1 | ENST00000587790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009235.hg.1 | ENST00000587790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009235.hg.1 | ENST00000587790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009235.hg.1 | ENST00000587790.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009235.hg.1 | NR_024021.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009235.hg.1 | NR_024021.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009235.hg.1 | NR_024021.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009235.hg.1 | NR_024021.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009235.hg.1 | NR_024021.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009235.hg.1 | NR_024021.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009235.hg.1 | ENST00000587689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009235.hg.1 | ENST00000587689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009235.hg.1 | ENST00000587689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009235.hg.1 | ENST00000587689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009235.hg.1 | ENST00000587689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009235.hg.1 | ENST00000587790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009235.hg.1 | ENST00000587790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009235.hg.1 | ENST00000587790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009235.hg.1 | ENST00000587790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009235.hg.1 | ENST00000587790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009235.hg.1 | ENST00000587790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009235.hg.1 | ENST00000587790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009235.hg.1 | ENST00000587790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009235.hg.1 | ENST00000587790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009235.hg.1 | ENST00000587790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009235.hg.1 | ENST00000587790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009235.hg.1 | ENST00000587790.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009235.hg.1 | NR_024021.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009235.hg.1 | NR_024021.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009235.hg.1 | NR_024021.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009235.hg.1 | NR_024021.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009235.hg.1 | NR_024021.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009235.hg.1 | NR_024021.2 | Refseq |

OACYLP
Genomic Map