Assay Details
| Gene Symbol: EIF4H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK297651.1 | - | 1 - 2 | 116 | 156 | |
| AK314749.1 | - | 1 - 2 | 173 | 156 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK297651.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000479338.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000479338.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000479338.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000479338.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000479338.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000479338.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000265753.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000353999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000479338.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000479338.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000479338.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000479338.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000479338.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000479338.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000489484.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | ENST00000495187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_022170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_031992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007998.hg.1 | NM_031992.1 | Refseq |

EIF4H
Genomic Map