Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized 100631378 | LOC100631378 | Anti-sense | Chr. 19 - 37823722 - 37855196 on NCBI Build GRCh38 | ||
| Downstream | WD repeat domain 87 | WDR87 | NYD-SP11 | 52.4kb | Anti-sense | Chr. 19 - 37884577 - 37906706 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map