| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000519176.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000519176.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000519176.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000519176.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000519176.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000519176.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000519176.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000519176.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000519353.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000519353.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000519353.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000519353.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000520774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000520774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000520774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000520774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000520774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000520774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000520774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000520774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000520774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000521525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000521525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000521525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000521525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000522422.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000522422.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000522422.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000522422.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000522422.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000522422.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000522422.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000522422.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000522422.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000522669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000522669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000522669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000522669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000522669.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000523020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000523020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000523020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000523020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000523020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000523020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000523020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
ENST00000523020.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007933.hg.1 |
ENST00000414645.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007933.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08007933.hg.1 |
TC08000577.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000519176.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000519176.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000519176.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000519176.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000519176.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000519176.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000519176.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000519176.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000519353.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000519353.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000519353.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000519353.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000520774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000520774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000520774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000520774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000520774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000520774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000520774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000520774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000520774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000521525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000521525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000521525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000521525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000522422.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000522422.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000522422.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000522422.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000522422.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000522422.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000522422.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000522422.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000522422.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000522669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000522669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000522669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000522669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000522669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000523020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000523020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000523020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000523020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000523020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000523020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000523020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
ENST00000523020.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008266.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800156839.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800156839.hg.1 |
ENST00000522669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800156839.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800156839.hg.1 |
ENST00000414645.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800156839.hg.1 |
ENST00000522669.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800156839.hg.1 |
NM_181787.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800156839.hg.1 |
TC0800008266.hg.1 |
Clariom D TC |