Assay Details
| Gene Symbol: ARHGAP40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164431.1 | NP_001157903.1 | 10 - 11 | 1368 | 99 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000287.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373346.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373346.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373346.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373346.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373346.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373346.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000373346.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000414158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000414158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000414158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | ENST00000414158.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004151.hg.1 | ENST00000373345.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004151.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004151.hg.1 | TC20000287.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373346.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373346.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373346.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373346.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373346.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373346.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000373346.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000414158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000414158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000414158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | ENST00000414158.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007327.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150747.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150747.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150747.hg.1 | NM_001164431.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150747.hg.1 | TC2000007327.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000150747.hg.1 | ENST00000243967.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150747.hg.1 | ENST00000373345.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150747.hg.1 | NM_001164431.1 | Refseq |

ARHGAP40
Genomic Map