Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | carnitine O-octanoyltransferase | CROT | COT | -76.0kb | Sense | Chr. 7 - 87345635 - 87399796 on NCBI Build GRCh38 |
| Downstream | ATP binding cassette subfamily B member 1 | ABCB1 |
ABC20, CD243, CLCS, GP170, MDR1, P-GP, PGY1 |
27.6kb | Anti-sense | Chr. 7 - 87503863 - 87713323 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map