Assay Details
| Gene Symbol: SASS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304829.1 | NP_001291758.1 | 9 - 10 | 1346 | 105 | |
| NM_194292.2 | NP_919268.1 | 10 - 11 | 1426 | 105 | ||
| XM_017000486.1 | XP_016855975.1 | 10 - 11 | 1192 | 105 | ||
| GenBank mRNA | AK302513.1 | - | 11 - 12 | 1176 | 105 | |
| AK307824.1 | - | 9 - 10 | 1212 | 105 | ||
| AL834265.1 | - | 6 - 7 | 689 | 105 | ||
| AY359522.1 | - | 10 - 11 | 1286 | 105 | ||
| BC101026.3 | - | 9 - 10 | 1139 | 40007432 | 105 | |
| BX641109.1 | - | 9 - 10 | 1337 | 105 | ||
| BX648406.1 | - | 10 - 11 | 1183 | 105 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002916.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01045311.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045311.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045311.hg.1 | ENST00000535161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045311.hg.1 | ENST00000539329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045311.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045311.hg.1 | TC01002916.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000432294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000432294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000432294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000432294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212164.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212164.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212164.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212164.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212164.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212164.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212164.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212164.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212164.hg.1 | TC0100015058.hg.1 | Clariom D TC |

SASS6
Genomic Map