Assay Details
| Gene Symbol: SASS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194292.2 | NP_919268.1 | 2 - 3 | 399 | 105 | |
| GenBank mRNA | AK302513.1 | - | 3 - 4 | 149 | 105 | |
| AY359522.1 | - | 2 - 3 | 259 | 105 | ||
| BC054889.1 | - | 2 - 3 | 262 | 105 | ||
| BX641109.1 | - | 2 - 3 | 218 | 105 | ||
| BX648406.1 | - | 2 - 3 | 155 | 105 | ||
| DC400869.1 | - | 2 - 3 | 399 | 105 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002916.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK302513.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000432294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000432294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045319.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045319.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045319.hg.1 | ENST00000535161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045319.hg.1 | ENST00000539329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045319.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045319.hg.1 | TC01002916.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000432294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000432294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212174.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212174.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212174.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212174.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212174.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212174.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212174.hg.1 | TC0100015058.hg.1 | Clariom D TC |

SASS6
Genomic Map