Assay Details
| Gene Symbol: SASS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304829.1 | NP_001291758.1 | 8 - 9 | 1256 | 147 | |
| NM_194292.2 | NP_919268.1 | 9 - 10 | 1336 | 147 | ||
| XM_017000486.1 | XP_016855975.1 | 9 - 10 | 1102 | 147 | ||
| GenBank mRNA | AK302513.1 | - | 10 - 11 | 1086 | 147 | |
| AK307824.1 | - | 8 - 9 | 1122 | 147 | ||
| AL834265.1 | - | 5 - 6 | 599 | 147 | ||
| AY359522.1 | - | 9 - 10 | 1196 | 147 | ||
| BC101026.3 | - | 8 - 9 | 1049 | 40007432 | 147 | |
| BX648406.1 | - | 9 - 10 | 1093 | 148 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002916.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01045312.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045312.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045312.hg.1 | ENST00000535161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045312.hg.1 | ENST00000539329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045312.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045312.hg.1 | TC01002916.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000432294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000432294.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000432294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000432294.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015058.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212165.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212165.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212165.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212165.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212165.hg.1 | ENST00000287482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212165.hg.1 | ENST00000462159.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100212165.hg.1 | NM_001304829.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212165.hg.1 | NM_194292.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100212165.hg.1 | TC0100015058.hg.1 | Clariom D TC |

SASS6
Genomic Map