Assay Details
| Gene Symbol: LOC100128922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF251047.1 | - | 1 | 738 | 147 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 5 member 9 | SLC5A9 | SGLT4 | -13.6kb | Anti-sense | Chr. 1 - 48221113 - 48248644 on NCBI Build GRCh38 |
| Downstream | ATP/GTP binding protein like 4 | AGBL4 | CCP6 | 269.7kb | Sense | Chr. 1 - 48520157 - 50024011 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00185885 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212069 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212056 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212048 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212035 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212078 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00185877 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00185882 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212073 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212062 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212080 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212031 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212067 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212060 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212066 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212061 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00185852 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212033 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00185880 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212079 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212059 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212049 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212063 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00185841 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212055 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212072 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15058529 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212064 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00185836 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00185848 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212034 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212040 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212065 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00185849 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212030 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212045 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212068 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00185883 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00185856 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212071 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20212054 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00185881 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P204989 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P204989 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 37576 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 24221 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 10249 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P204989 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7915953 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7915954 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7915952 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7915951 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 224362_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11762958_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11762958_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014126.hg.1 | ENST00000624985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014126.hg.1 | ENST00000624985.1 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 224362_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 224362_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224362_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224362_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224362_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224362_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224362_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224362_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224362_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224362_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224362_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224362_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224362_at5 | - | - |
| Human Genome U133 Set | 224362_at | - | - |
| Human Genome U133 Set | 224362_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 234118 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3267831 | AF251047.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1826215 | AF251047.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1826215 | AF251047.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1826215 | AF251047.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_103702 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_103702 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1826215 | AF251047.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1826215 | AF251047.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1826215 | AF251047.1 | human_transcriptome |

LOC100128922
Genomic Map