Assay Details
| Gene Symbol: BEND4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159547.1 | NP_001153019.1 | 3 - 4 | 1639 | 88 | |
| NM_207406.3 | NP_997289.2 | 3 - 4 | 1639 | 88 | ||
| XM_017008185.1 | XP_016863674.1 | 2 - 3 | 1296 | 88 | ||
| GenBank mRNA | AB126828.1 | - | 3 - 4 | 1639 | 88 | |
| AB126829.1 | - | 3 - 4 | 1639 | 88 | ||
| AK125953.1 | - | 3 - 4 | 825 | 88 | ||
| BC128592.1 | - | 3 - 4 | 724 | 40127911 | 88 | |
| BC128593.1 | - | 3 - 4 | 724 | 40127915 | 88 | |
More Information
Set Membership: |
|

BEND4
Genomic Map