| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001874.hg.1 |
ENST00000267889.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001874.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001874.hg.1 |
TC15000281.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001873.hg.1 |
ENST00000267889.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001873.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15001873.hg.1 |
TC15000281.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1500181786.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500181786.hg.1 |
TC1500010717.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500181786.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500181785.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500181785.hg.1 |
TC1500010717.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500181785.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500181787.hg.1 |
NM_033510.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500181787.hg.1 |
TC1500010717.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500181787.hg.1 |
NM_033510.2 |
Refseq |