Assay Details
| Gene Symbol: EXOC3L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077594.1 | NP_001071062.1 | 5 - 6 | 1463 | 144 | |
| XM_011537323.2 | XP_011535625.1 | 6 - 7 | 2138 | 144 | ||
| XM_011537324.2 | XP_011535626.1 | 6 - 7 | 2140 | 144 | ||
| XM_011537325.2 | XP_011535627.1 | 7 - 8 | 1894 | 144 | ||
| XM_011537327.2 | XP_011535629.1 | 6 - 7 | 1593 | 144 | ||
| XM_011537328.2 | XP_011535630.1 | 7 - 8 | 1625 | 144 | ||
| XM_011537329.2 | XP_011535631.1 | 7 - 8 | 2033 | 144 | ||
| XM_011537330.2 | XP_011535632.1 | 8 - 9 | 1814 | 144 | ||
| XM_011537331.1 | XP_011535633.1 | 7 - 8 | 1609 | 144 | ||
| XM_011537332.2 | XP_011535634.1 | 6 - 7 | 1838 | 144 | ||
| XM_011537333.2 | XP_011535635.1 | 7 - 8 | 1870 | 144 | ||
| XM_011537334.1 | XP_011535636.1 | 5 - 6 | 1821 | 144 | ||
| GenBank mRNA | BC113655.1 | - | 5 - 6 | 1463 | 8322707 | 144 |
| BC117375.1 | - | 5 - 6 | 1463 | 40125926 | 144 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000793.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14008550.hg.1 | ENST00000380069.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14008550.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14008550.hg.1 | TC14000793.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000559116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000559116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000559116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000559661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000559661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000559661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000559661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000559661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000559661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000559661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000559661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000559693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000559693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000559693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000559693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000559693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000559693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000559693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000559693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000559116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000559116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000559116.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000559661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000559661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000559661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000559661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000559661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000559661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000559661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000559661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000559693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000559693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000559693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000559693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000559693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000559693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000559693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000559693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008380.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400159757.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400159757.hg.1 | ENST00000559661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400159757.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400159757.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400159757.hg.1 | ENST00000380069.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400159757.hg.1 | ENST00000559661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400159757.hg.1 | ENST00000560925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400159757.hg.1 | NM_001077594.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400159757.hg.1 | TC1400008380.hg.1 | Clariom D TC |

EXOC3L4
Genomic Map