Assay Details
| Gene Symbol: ABHD17C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021214.1 | NP_067037.1 | 1 - 2 | 705 | 142 | |
| GenBank mRNA | AK298606.1 | - | 1 - 2 | 291 | 142 | |
| AL390079.1 | - | 1 - 2 | 326 | 142 | ||
| BC059401.1 | - | 1 - 2 | 675 | 30346383 | 142 | |
| BC115003.1 | - | 1 - 2 | 705 | 7503983 | 142 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15000751.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15006821.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15006821.hg.1 | ENST00000559506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15006821.hg.1 | ENST00000560609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15006821.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15006821.hg.1 | TC15000751.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000559506.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000559506.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000559506.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000559506.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000559506.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000559506.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000559506.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000559506.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000560126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000560126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000560126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000560126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000560126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000560609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000560609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000560609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000560609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000560609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | ENST00000560609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008043.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15006818.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15006818.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15006818.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15006818.hg.1 | TC15000751.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15006820.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15006820.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15006820.hg.1 | ENST00000559506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15006820.hg.1 | ENST00000560609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15006820.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15006820.hg.1 | TC15000751.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15006817.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15006817.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15006817.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15006817.hg.1 | TC15000751.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1500157830.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157830.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157830.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500157830.hg.1 | TC1500008043.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500157830.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157830.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157830.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500157836.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157836.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157836.hg.1 | ENST00000559506.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157836.hg.1 | ENST00000560126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157836.hg.1 | ENST00000560609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157836.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500157836.hg.1 | TC1500008043.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500157836.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157836.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157836.hg.1 | ENST00000559506.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157836.hg.1 | ENST00000560126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157836.hg.1 | ENST00000560609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157836.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500157832.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157832.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157832.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500157832.hg.1 | TC1500008043.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500157832.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157832.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157832.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000559506.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000559506.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000559506.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000559506.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000559506.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000559506.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000559506.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000559506.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000560126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000560126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000560126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000560126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000560126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000560609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000560609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000560609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000560609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000560609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | ENST00000560609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008043.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500157831.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157831.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157831.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500157831.hg.1 | TC1500008043.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500157831.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157831.hg.1 | ENST00000558464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157831.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500157837.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157837.hg.1 | ENST00000559506.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157837.hg.1 | ENST00000560126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157837.hg.1 | ENST00000560609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157837.hg.1 | NM_021214.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500157837.hg.1 | TC1500008043.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500157837.hg.1 | ENST00000258884.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157837.hg.1 | ENST00000559506.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157837.hg.1 | ENST00000560126.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157837.hg.1 | ENST00000560609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500157837.hg.1 | NM_021214.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 101408 | - | - |

ABHD17C
Genomic Map