Assay Details
| Gene Symbol: SEC14L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011530164.2 | XP_011528466.1 | 3 - 4 | 453 | 116 | |
| GenBank mRNA | AK092002.1 | - | 4 - 5 | 322 | 116 | |
More Information
Set Membership: |
|

SEC14L4
Genomic Map