Assay Details
| Gene Symbol: FBXW4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001323541.1 | NP_001310470.1 | 4 - 5 | 465 | 162 | |
| NM_022039.3 | NP_071322.1 | 4 - 5 | 1015 | 162 | ||
| XM_005270053.2 | XP_005270110.2 | 4 - 5 | 1066 | 162 | ||
| XM_017016570.1 | XP_016872059.1 | 4 - 5 | 1066 | 162 | ||
| GenBank mRNA | AF281859.1 | - | 4 - 5 | 1288 | 162 | |
| AK314735.1 | - | 4 - 5 | 960 | 162 | ||
| BC007380.2 | - | 2 - 3 | 314 | 162 | ||
| BC063415.1 | - | 4 - 5 | 782 | 5223794 | 162 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001609.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10020859.hg.1 | ENST00000331272.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020859.hg.1 | ENST00000389046.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020859.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020859.hg.1 | TC10001609.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000482428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000482428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000482428.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000489578.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000489578.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | ENST00000489578.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020860.hg.1 | ENST00000331272.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020860.hg.1 | ENST00000389046.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020860.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10020860.hg.1 | TC10001609.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006836.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006836.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006836.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006836.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006836.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006836.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006836.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006836.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006836.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006836.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179655.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179655.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179655.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179655.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179655.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179655.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179655.hg.1 | TC1000011652.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC2200006836.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006836.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006836.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006836.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006836.hg.1 | ENST00000426721.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006836.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006836.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006836.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006836.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006836.hg.1 | NR_033408.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179652.hg.1 | TC1000011652.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000331272.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000457105.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000470093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000482428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000482428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000482428.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000489578.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000489578.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | ENST00000489578.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011652.hg.1 | NM_022039.3 | Refseq |

FBXW4
Genomic Map