Assay Details
| Gene Symbol: USP17L7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256869.1 | NP_001243798.1 | 1 | 1533 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

USP17L7| Gene Symbol: USP17L7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256869.1 | NP_001243798.1 | 1 | 1533 | 124 | |
Set Membership: |
|