Assay Details
| Gene Symbol: LYSMD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286812.1 | NP_001273741.1 | 1 - 2 | 126 | 98 | |
| NM_198273.2 | NP_938014.1 | 1 - 2 | 126 | 98 | ||
| GenBank mRNA | AK291478.1 | - | 1 - 2 | 123 | 98 | |
| AL832721.1 | - | 1 - 2 | 107 | 98 | ||
| BC058027.1 | - | 1 - 2 | 126 | 98 | ||
| BP873090.1 | - | 1 - 2 | 127 | 98 | ||
| BX537972.1 | - | 1 - 2 | 96 | 98 | ||
| DN994993.1 | - | 1 - 2 | 135 | 98 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001586.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK291478.1,BC058027.1,NM_001286812.1,NM_198273.2 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05021589.hg.1 | ENST00000315948.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05021589.hg.1 | ENST00000500869.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05021589.hg.1 | ENST00000509384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05021589.hg.1 | ENST00000554351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05021589.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05021589.hg.1 | TC05001586.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05021587.hg.1 | ENST00000315948.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05021587.hg.1 | ENST00000500869.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05021587.hg.1 | ENST00000509384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05021587.hg.1 | ENST00000554351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05021587.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05021587.hg.1 | TC05001586.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05021590.hg.1 | ENST00000315948.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05021590.hg.1 | ENST00000500869.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05021590.hg.1 | ENST00000509384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05021590.hg.1 | ENST00000554351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05021590.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05021590.hg.1 | TC05001586.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0500179166.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179166.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179166.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179166.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179166.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179166.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179166.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179166.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179166.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179166.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179166.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179166.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179166.hg.1 | TC0500011435.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500179169.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179169.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179169.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179169.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179169.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179169.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179169.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179169.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179169.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179169.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179169.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179169.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179169.hg.1 | TC0500011435.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500179170.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179170.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179170.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179170.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179170.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179170.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179170.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179170.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179170.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179170.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179170.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179170.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179170.hg.1 | TC0500011435.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_198273.2 | Refseq |

LYSMD3
Genomic Map