Assay Details
| Gene Symbol: LYSMD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286812.1 | NP_001273741.1 | 2 - 3 | 407 | 86 | |
| GenBank mRNA | BC058027.1 | - | 2 - 3 | 407 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179159.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179159.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179159.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179159.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179159.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179159.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179159.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179159.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179159.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179159.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179159.hg.1 | TC0500011435.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500179161.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179161.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179161.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179161.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179161.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179161.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179161.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179161.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179161.hg.1 | TC0500011435.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500179157.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179157.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179157.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179157.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179157.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179157.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179157.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179157.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179157.hg.1 | TC0500011435.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500179158.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179158.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179158.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179158.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179158.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179158.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179158.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179158.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179158.hg.1 | TC0500011435.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500179162.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179162.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179162.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179162.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179162.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179162.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179162.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179162.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179162.hg.1 | TC0500011435.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000453259.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011435.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179160.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179160.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179160.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179160.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179160.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179160.hg.1 | ENST00000315948.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179160.hg.1 | ENST00000500869.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179160.hg.1 | ENST00000509384.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179160.hg.1 | NM_001286812.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179160.hg.1 | NM_198273.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179160.hg.1 | TC0500011435.hg.1 | Clariom D TC |

LYSMD3
Genomic Map