Assay Details
| Gene Symbol: DNAJC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC036571.1 | - | 4 - 5 | 506 | 4793909 | 144 |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000395949.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000395949.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000395949.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000395949.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000524747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000524747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000524747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000524747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000524747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000524747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000525511.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000525511.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000525511.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000525511.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000525511.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000525511.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000526529.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000526529.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000526529.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000526529.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000526529.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000526529.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000526529.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000526529.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000527601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000527601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000527601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000527601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000527601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000527731.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000527731.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000527731.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000527731.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000527731.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000527731.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000527731.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000529086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000529086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000529086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000529086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000529086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000529086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000529086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000529086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000529086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000530125.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000530125.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000530125.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000532385.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000532385.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000532385.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000532385.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000532385.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | ENST00000532385.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100150513.hg.1 | ENST00000524747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150513.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150513.hg.1 | ENST00000524747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150513.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150513.hg.1 | TC1100007184.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000395949.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000395949.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000395949.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000395949.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000524747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000524747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000524747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000524747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000524747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000524747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000525511.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000525511.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000525511.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000525511.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000525511.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000525511.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000526529.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000526529.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000526529.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000526529.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000526529.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000526529.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000526529.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000526529.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000527601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000527601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000527601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000527601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000527601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000527731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000527731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000527731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000527731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000527731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000527731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000527731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000529086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000529086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000529086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000529086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000529086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000529086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000529086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000529086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000529086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000530125.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000530125.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000530125.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000532385.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000532385.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000532385.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000532385.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000532385.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | ENST00000532385.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007184.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100150515.hg.1 | ENST00000524747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150515.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150515.hg.1 | ENST00000527731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150515.hg.1 | ENST00000524747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150515.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150515.hg.1 | ENST00000527731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150515.hg.1 | TC1100007184.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100150516.hg.1 | ENST00000524747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150516.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150516.hg.1 | ENST00000524747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150516.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150516.hg.1 | TC1100007184.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100150514.hg.1 | ENST00000395949.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150514.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150514.hg.1 | ENST00000524747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150514.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150514.hg.1 | ENST00000527731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150514.hg.1 | ENST00000530125.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150514.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100150514.hg.1 | ENST00000395949.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150514.hg.1 | ENST00000465995.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150514.hg.1 | ENST00000524747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150514.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150514.hg.1 | ENST00000527731.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150514.hg.1 | ENST00000530125.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150514.hg.1 | NM_181706.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100150514.hg.1 | TC1100007184.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100150517.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150517.hg.1 | ENST00000526042.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150517.hg.1 | TC1100007184.hg.1 | Clariom D TC |

DNAJC24
Genomic Map