Assay Details
| Gene Symbol: GANC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198141.2 | NP_937784.2 | 12 - 13 | 1733 | 144 | |
| GenBank mRNA | AF525397.1 | - | 8 - 9 | 986 | 144 | |
| AF545045.1 | - | 12 - 13 | 1427 | 144 | ||
| AF545046.1 | - | 12 - 13 | 1427 | 144 | ||
| AK074037.1 | - | 12 - 13 | 1625 | 144 | ||
| BC059406.1 | - | 12 - 13 | 1605 | 30330796 | 144 | |
| BC093833.1 | - | 12 - 13 | 1450 | 7939678 | 144 | |
| BC112051.1 | - | 12 - 13 | 1450 | 8327519 | 144 | |
More Information
Set Membership: |
|

GANC
Genomic Map