Assay Details
| Gene Symbol: KCTD20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286580.1 | NP_001273509.1 | 4 - 5 | 773 | 149 | |
| NM_173562.4 | NP_775833.2 | 5 - 6 | 1047 | 149 | ||
| XM_005248940.4 | XP_005248997.1 | 6 - 7 | 902 | 149 | ||
| XM_006715023.2 | XP_006715086.1 | 5 - 6 | 820 | 149 | ||
| XM_011514398.2 | XP_011512700.1 | 6 - 7 | 898 | 149 | ||
| GenBank mRNA | AK302861.1 | - | 4 - 5 | 504 | 149 | |
| AK304377.1 | - | 5 - 6 | 768 | 149 | ||
| AL832381.1 | - | 4 - 5 | 559 | 149 | ||
| AY305862.1 | - | 5 - 6 | 1047 | 149 | ||
More Information
Updated Mapping Information
Set Membership: |
|

KCTD20
Genomic Map