| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409948.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409948.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409948.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000409948.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000458251.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000458251.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000458251.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000462655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000462655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000462655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000462655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000462655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000498662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
ENST00000498662.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027673.hg.1 |
ENST00000389942.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027673.hg.1 |
ENST00000409806.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027673.hg.1 |
ENST00000416573.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027673.hg.1 |
ENST00000542614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027673.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027673.hg.1 |
TC06002209.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027669.hg.1 |
ENST00000389942.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027669.hg.1 |
ENST00000409806.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027669.hg.1 |
ENST00000409948.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027669.hg.1 |
ENST00000416573.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027669.hg.1 |
ENST00000542614.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027669.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06027669.hg.1 |
TC06002209.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409948.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409948.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409948.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000409948.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000458251.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000458251.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000458251.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000462655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000462655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000462655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000462655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000462655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000498662.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
ENST00000498662.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013519.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600193292.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600193292.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600193292.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600193292.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600193292.hg.1 |
TC0600013519.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600193288.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600193288.hg.1 |
ENST00000409948.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600193288.hg.1 |
ENST00000462655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600193288.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600193288.hg.1 |
ENST00000409806.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600193288.hg.1 |
ENST00000409948.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600193288.hg.1 |
ENST00000462655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600193288.hg.1 |
NM_207360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600193288.hg.1 |
TC0600013519.hg.1 |
Clariom D TC |