Assay Details
| Gene Symbol: SEC14L6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017028933.1 | XP_016884422.1 | 10 - 11 | 943 | 177 | |
| XM_017028934.1 | XP_016884423.1 | 10 - 11 | 1157 | 177 | ||
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | ENST00000437871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | ENST00000437871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | ENST00000437871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | ENST00000437871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | ENST00000437871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | ENST00000437871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | ENST00000437871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | NM_001193336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | NM_001193336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | NM_001193336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | NM_001193336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | NM_001193336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | NM_001193336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | NM_001193336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | NM_001193336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | NM_001193336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | NM_001193336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | NM_001193336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | NM_001193336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008445.hg.1 | NM_001193336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | ENST00000402034.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | ENST00000437871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | ENST00000437871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | ENST00000437871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | ENST00000437871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | ENST00000437871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | ENST00000437871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | ENST00000437871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | NM_001193336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | NM_001193336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | NM_001193336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | NM_001193336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | NM_001193336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | NM_001193336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | NM_001193336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | NM_001193336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | NM_001193336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | NM_001193336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | NM_001193336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | NM_001193336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008445.hg.1 | NM_001193336.3 | Refseq |

SEC14L6
Genomic Map