| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006530.hg.1 |
ENST00000582054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006530.hg.1 |
ENST00000582054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006530.hg.1 |
ENST00000582054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006530.hg.1 |
ENST00000582054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006530.hg.1 |
ENST00000582054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006530.hg.1 |
NR_102696.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006530.hg.1 |
NR_102696.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006530.hg.1 |
NR_102696.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006530.hg.1 |
NR_102696.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800006530.hg.1 |
NR_102696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006530.hg.1 |
ENST00000582054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006530.hg.1 |
ENST00000582054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006530.hg.1 |
ENST00000582054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006530.hg.1 |
ENST00000582054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006530.hg.1 |
ENST00000582054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800006530.hg.1 |
NR_102696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006530.hg.1 |
NR_102696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006530.hg.1 |
NR_102696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006530.hg.1 |
NR_102696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800006530.hg.1 |
NR_102696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800144752.hg.1 |
ENST00000582054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800144752.hg.1 |
NR_102696.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800144752.hg.1 |
TC1800006530.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800144752.hg.1 |
ENST00000582054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800144752.hg.1 |
NR_102696.1 |
Refseq |