Assay Details
| Gene Symbol: CCDC162P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC107113.2 | - | 3 - 4 | 463 | 40022743 | 154 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000422819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000422819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000422819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000422819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000422819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000422819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000422819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000422819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000422819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000429614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000429614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000429614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000429614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000429614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000429614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000429614.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000514758.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000514758.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000514758.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000514758.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000514758.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000615766.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000615766.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000615766.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000615766.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000615766.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000615766.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000615766.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000615766.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | ENST00000615766.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | NR_028595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | NR_028595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | NR_028595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | NR_028595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | NR_028595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | NR_028595.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014166.hg.1 | NR_028595.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600199290.hg.1 | ENST00000422819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600199290.hg.1 | ENST00000429614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600199290.hg.1 | ENST00000615766.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600199290.hg.1 | NR_028595.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600199290.hg.1 | ENST00000422819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600199290.hg.1 | ENST00000429614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600199290.hg.1 | ENST00000615766.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600199290.hg.1 | NR_028595.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600199290.hg.1 | TC0600014166.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000422819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000422819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000422819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000422819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000422819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000422819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000422819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000422819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000422819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000429614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000429614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000429614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000429614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000429614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000429614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000429614.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000514758.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000514758.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000514758.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000514758.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000514758.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000615766.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000615766.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000615766.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000615766.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000615766.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000615766.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000615766.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000615766.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | ENST00000615766.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | NR_028595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | NR_028595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | NR_028595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | NR_028595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | NR_028595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | NR_028595.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014166.hg.1 | NR_028595.1 | Refseq |

CCDC162P
Genomic Map