Assay Details
| Gene Symbol: SUCLA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017020809.1 | XP_016876298.1 | 3 - 4 | 416 | 133 | |
| GenBank mRNA | AK302311.1 | - | 3 - 4 | 391 | 133 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK302311.1, XM_017020809.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200011997.hg.1 | ENST00000458157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011997.hg.1 | ENST00000458157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011997.hg.1 | ENST00000458157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000378654.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000378654.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000378654.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000378654.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000378654.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000378654.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000378654.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000378654.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000378654.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000378654.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000378654.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000378654.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000378654.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000378654.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000378654.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000378654.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000378654.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000433022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000433022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000433022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000433022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000433022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000433022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000434484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000434484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000434484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000434484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000434484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000434484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000434484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000434484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000434484.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000467222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000467222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000467222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000467222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000467222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000467222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000467222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000467222.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000470760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000470760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000470760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000470760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000470760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000470760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000481279.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000481279.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000481279.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000484941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000484941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000484941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000493152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000493152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000493152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000493152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000493152.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000497202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000497202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000497202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000497202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000497202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000497202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000497202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | ENST00000497202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | NM_003850.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | NM_003850.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | NM_003850.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | NM_003850.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | NM_003850.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | NM_003850.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | NM_003850.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | NM_003850.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | NM_003850.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | NM_003850.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | NM_003850.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | NM_003850.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | NM_003850.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | NM_003850.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | NM_003850.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | NM_003850.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008873.hg.1 | NM_003850.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011997.hg.1 | ENST00000458157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011997.hg.1 | ENST00000458157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011997.hg.1 | ENST00000458157.1 | Ensembl |

SUCLA2
Genomic Map