Assay Details
| Gene Symbol: GSTA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BG573805.1 | - | 56 | 81 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600012056.hg.1 | ENST00000412182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012056.hg.1 | ENST00000412182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012056.hg.1 | ENST00000412182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012056.hg.1 | ENST00000412182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012056.hg.1 | ENST00000412182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012054.hg.1 | ENST00000424144.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012054.hg.1 | ENST00000424144.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012054.hg.1 | ENST00000424144.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012054.hg.1 | ENST00000424144.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012054.hg.1 | ENST00000424144.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014298.hg.1 | ENST00000452866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014298.hg.1 | ENST00000452866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014298.hg.1 | ENST00000452866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014298.hg.1 | ENST00000452866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014298.hg.1 | ENST00000452866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014298.hg.1 | ENST00000452866.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | ENST00000211122.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | ENST00000211122.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | ENST00000211122.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | ENST00000211122.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | ENST00000211122.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | ENST00000211122.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | ENST00000211122.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | ENST00000211122.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | ENST00000211122.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | ENST00000370968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | ENST00000370968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | ENST00000370968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | ENST00000370968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | ENST00000370968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | ENST00000370968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | ENST00000370968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | ENST00000370968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | ENST00000431899.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | ENST00000431899.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | ENST00000431899.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | ENST00000431899.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | NM_000847.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | NM_000847.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | NM_000847.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | NM_000847.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | NM_000847.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | NM_000847.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | NM_000847.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | NM_000847.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014299.hg.1 | NM_000847.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012056.hg.1 | ENST00000412182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012056.hg.1 | ENST00000412182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012056.hg.1 | ENST00000412182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012056.hg.1 | ENST00000412182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012056.hg.1 | ENST00000412182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | ENST00000211122.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | ENST00000211122.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | ENST00000211122.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | ENST00000211122.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | ENST00000211122.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | ENST00000211122.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | ENST00000211122.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | ENST00000211122.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | ENST00000211122.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | ENST00000370968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | ENST00000370968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | ENST00000370968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | ENST00000370968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | ENST00000370968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | ENST00000370968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | ENST00000370968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | ENST00000370968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | ENST00000431899.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | ENST00000431899.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | ENST00000431899.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | ENST00000431899.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | NM_000847.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | NM_000847.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | NM_000847.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | NM_000847.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | NM_000847.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | NM_000847.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | NM_000847.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | NM_000847.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014299.hg.1 | NM_000847.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012054.hg.1 | ENST00000424144.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012054.hg.1 | ENST00000424144.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012054.hg.1 | ENST00000424144.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012054.hg.1 | ENST00000424144.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012054.hg.1 | ENST00000424144.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014298.hg.1 | ENST00000452866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014298.hg.1 | ENST00000452866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014298.hg.1 | ENST00000452866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014298.hg.1 | ENST00000452866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014298.hg.1 | ENST00000452866.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014298.hg.1 | ENST00000452866.1 | Ensembl |

GSTA3
Genomic Map