Assay Details
| Gene Symbol: QPCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012413.3 | NP_036545.1 | 4 - 5 | 847 | 143 | |
| GenBank mRNA | AK290605.1 | - | 4 - 5 | 829 | 143 | |
| BC020456.1 | - | 4 - 5 | 748 | 143 | ||
| BC036721.1 | - | 4 - 5 | 726 | 5224222 | 143 | |
| BC047756.2 | - | 4 - 5 | 824 | 6058335 | 143 | |
| X67731.1 | - | 3 - 4 | 529 | 143 | ||
| X71125.1 | - | 4 - 5 | 736 | 143 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000237.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000444022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000444022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000444022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000444022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000444022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000470075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000470075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000470075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000470075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000470075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000480050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000480050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000480050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | ENST00000480050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003546.hg.1 | ENST00000338415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003546.hg.1 | ENST00000537448.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003546.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003546.hg.1 | TC02000237.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000404976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000444022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000444022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000444022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000444022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000444022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000469098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000470075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000470075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000470075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000470075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000470075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000480050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000480050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000480050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | ENST00000480050.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007261.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150285.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150285.hg.1 | ENST00000404976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150285.hg.1 | ENST00000444022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150285.hg.1 | ENST00000469098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150285.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150285.hg.1 | ENST00000338415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150285.hg.1 | ENST00000404976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150285.hg.1 | ENST00000444022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150285.hg.1 | ENST00000469098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150285.hg.1 | NM_012413.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150285.hg.1 | TC0200007261.hg.1 | Clariom D TC |

QPCT
Genomic Map