Assay Details
| Gene Symbol: PRNT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024267.1 | 1 - 2 | 524 | 105 | ||
| GenBank mRNA | AJ427540.1 | - | 1 - 2 | 524 | 105 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ427540.1 transcripts.
Set Membership: |
|

PRNT
Genomic Map