Assay Details
| Gene Symbol: YJEFN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190328.1 | NP_001177257.1 | 1 - 2 | 138 | 81 | |
| GenBank mRNA | AK126916.1 | - | 1 - 2 | 101 | 81 | |
| BI520231.1 | - | 1 - 2 | 138 | 81 | ||
| DQ778079.1 | - | 1 - 2 | 66 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK126916.1 transcripts.
Set Membership: |
|

YJEFN3
Genomic Map