| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000436027.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000436027.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000436027.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000436027.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000436027.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000436027.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000436027.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000436027.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000458210.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000458210.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000458210.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000458210.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000458210.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000458210.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000458210.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000458210.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000458210.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000514277.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000514277.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000514277.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000514277.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000514277.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000514277.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000514277.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000514277.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
ENST00000514277.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
NM_001190328.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
NM_001190328.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
NM_001190328.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
NM_001190328.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
NM_001190328.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
NM_001190328.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
NM_001190328.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
NM_001190328.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
NM_198537.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
NM_198537.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
NM_198537.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
NM_198537.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
NM_198537.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
NM_198537.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
NM_198537.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
NM_198537.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011689.hg.1 |
NM_198537.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000436027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000436027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000436027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000436027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000436027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000436027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000436027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000436027.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000458210.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000458210.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000458210.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000458210.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000458210.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000458210.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000458210.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000458210.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000458210.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000514277.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000514277.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000514277.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000514277.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000514277.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000514277.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000514277.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000514277.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
ENST00000514277.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
NM_001190328.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
NM_001190328.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
NM_001190328.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
NM_001190328.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
NM_001190328.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
NM_001190328.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
NM_001190328.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
NM_001190328.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
NM_198537.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
NM_198537.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
NM_198537.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
NM_198537.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
NM_198537.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
NM_198537.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
NM_198537.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
NM_198537.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011689.hg.1 |
NM_198537.3 |
Refseq |