Assay Details
| Gene Symbol: LOC400627 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK127919.1 | - | 1 - 2 | 427 | 75 | |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10491055 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17024651 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31077673 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31077661 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31077669 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10491056 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10491058 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31077666 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10491062 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10491060 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31077671 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31077674 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 807851 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 617285 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 288091 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 12615 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03299195 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P40919604 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | XR_041190_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | XR_041189_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1565905_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1565903_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012003.hg.1 | ENST00000573602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012003.hg.1 | ENST00000573602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012003.hg.1 | ENST00000573602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012003.hg.1 | ENST00000576234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012003.hg.1 | ENST00000576234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012003.hg.1 | ENST00000576234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012003.hg.1 | ENST00000576234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012003.hg.1 | ENST00000573602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012003.hg.1 | ENST00000573602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012003.hg.1 | ENST00000573602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012003.hg.1 | ENST00000576234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012003.hg.1 | ENST00000576234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012003.hg.1 | ENST00000576234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012003.hg.1 | ENST00000576234.1 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 1565903_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1565905_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1565903_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1565905_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565905_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565905_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565905_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565905_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565905_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565905_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565905_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565905_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565905_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565905_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565905_at11 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 339143 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1684193 | AK127919.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1805753 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1805753 | - | - |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1805753 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1684193 | AK127919.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1684193 | AK127919.1 | human_transcriptome |

LOC400627
Genomic Map