Assay Details
Gene Symbol: UBE2E4P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110506.1 | 1 | 437 | 151 | ||
GenBank mRNA | AF136176.1 | - | 1 | 437 | 151 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602230 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602178 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602284 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307192 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307146 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602217 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602189 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307138 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307155 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307142 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307102 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307107 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602206 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307148 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602190 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602196 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307124 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602254 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307176 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307111 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307131 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602187 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307189 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307141 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307216 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307167 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307219 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307215 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307211 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602259 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307182 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307172 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307196 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307218 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307204 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307133 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307109 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602294 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307171 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307129 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307132 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602277 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602213 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307213 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602257 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307190 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602252 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602285 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602261 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17320747 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307181 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602239 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602229 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307110 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307170 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307134 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307143 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307220 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307209 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307197 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307214 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307199 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602216 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307212 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307104 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602172 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602235 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602234 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307195 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602267 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307120 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307168 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602245 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307205 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307144 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602251 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307125 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307112 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602249 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307156 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307188 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602214 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602233 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307147 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307201 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307145 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307187 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602185 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602250 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307194 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307203 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602202 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602174 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602186 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602201 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32307210 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11602193 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8046693 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8046694 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8046690 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8046689 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8046691 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8166126 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8046692 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8166124 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8046685 | - | - |
Affymetrix Human Clariom D Assay | PSR0X00145599.hg.1 | NR_110506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00145599.hg.1 | NR_110506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00145599.hg.1 | TC0X00006666.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00145598.hg.1 | ENST00000411687.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00145598.hg.1 | NR_110506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00145598.hg.1 | ENST00000411687.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00145598.hg.1 | NR_110506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00145598.hg.1 | TC0X00006666.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00145597.hg.1 | NR_110506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00145597.hg.1 | NR_110506.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00145597.hg.1 | TC0X00006666.hg.1 | Clariom D TC |
Human Genome U95 Set | 63933_s_at | - | - |
Human Genome U95 Set | 63933_PM_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 165922 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 188548 | - | - |
