Assay Details
| Gene Symbol: PGAM1P4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC171813.1 | - | 1 | 198 | 9054467 | 74 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC171813.1 transcripts.
Set Membership: |
|

PGAM1P4
Genomic Map