| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03027563.hg.1 |
ENST00000462497.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03027563.hg.1 |
ENST00000466893.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03027563.hg.1 |
ENST00000474795.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03027563.hg.1 |
ENST00000475371.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03027563.hg.1 |
ENST00000479018.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03027563.hg.1 |
ENST00000490916.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03027563.hg.1 |
ENST00000495542.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03027563.hg.1 |
ENST00000498655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03027563.hg.1 |
NR_038281.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03027563.hg.1 |
NR_038282.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03027563.hg.1 |
NR_038283.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03027563.hg.1 |
TC03001514.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000462497.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000462497.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000466893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000466893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000474795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000474795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000475371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000475371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000479018.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000479018.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000479588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000490916.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000490916.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000495542.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000495542.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000498655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000498655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
NR_038281.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
NR_038282.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
NR_038283.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000462497.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000462497.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000466893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000466893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000474795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000474795.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000475371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000475371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000479018.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000479018.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000479588.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000490916.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000490916.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000495542.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000495542.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000498655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
ENST00000498655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
NR_038281.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
NR_038282.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
NR_038283.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300185160.hg.1 |
TC0300011360.hg.1 |
Clariom D TC |