Assay Details
| Gene Symbol: SLC16A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017024282.1 | XP_016879771.1 | 3 - 4 | 987 | 125 | |
| GenBank mRNA | BC143369.1 | - | 1 - 2 | 675 | 125 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000308009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000308009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000308009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000308009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000308009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000308009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000308009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000308009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000308009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000447225.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000447225.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000447225.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000447225.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000447225.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000447225.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000447225.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000447225.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000447225.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000573338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000573338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000573338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000573338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000573338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000573338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000574600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000574600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | ENST00000574600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | NM_153357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | NM_153357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | NM_153357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | NM_153357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | NM_153357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | NM_153357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | NM_153357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | NM_153357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009610.hg.1 | NM_153357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000308009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000308009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000308009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000308009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000308009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000308009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000308009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000308009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000308009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000447225.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000447225.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000447225.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000447225.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000447225.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000447225.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000447225.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000447225.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000447225.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000573338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000573338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000573338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000573338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000573338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000573338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000574600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000574600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | ENST00000574600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | NM_153357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | NM_153357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | NM_153357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | NM_153357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | NM_153357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | NM_153357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | NM_153357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | NM_153357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009610.hg.1 | NM_153357.1 | Refseq |

SLC16A11
Genomic Map