Assay Details
| Gene Symbol: SLFNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017000573.1 | XP_016856062.1 | 3 - 4 | 1958 | 91 | |
| GenBank mRNA | BC037879.1 | - | 2 - 3 | 2031 | 5274032 | 91 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000302946.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000302946.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000302946.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000302946.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000302946.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000302946.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000302946.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000302946.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000372611.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000372611.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000372611.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000372611.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000372611.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000372611.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000372611.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000372611.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000372613.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000372613.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000372613.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000372613.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000372613.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000372613.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000372613.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000372613.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000372613.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000372613.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000372613.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000439569.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000439569.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000439569.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000439569.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000439569.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000439569.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000439569.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000439569.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000439569.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | ENST00000439569.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_001168247.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_001168247.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_001168247.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_001168247.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_001168247.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_001168247.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_001168247.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_001168247.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_001168247.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_001168247.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_001300859.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_001300859.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_001300859.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_001300859.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_001300859.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_001300859.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_001300859.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_001300859.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_144990.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_144990.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_144990.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_144990.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_144990.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_144990.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_144990.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013853.hg.1 | NM_144990.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100203557.hg.1 | ENST00000302946.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203557.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203557.hg.1 | ENST00000372611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203557.hg.1 | ENST00000372613.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203557.hg.1 | ENST00000439569.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203557.hg.1 | NM_001168247.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100203557.hg.1 | NM_001300859.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100203557.hg.1 | NM_144990.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100203557.hg.1 | ENST00000302946.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203557.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203557.hg.1 | ENST00000372611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203557.hg.1 | ENST00000372613.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203557.hg.1 | ENST00000439569.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203557.hg.1 | NM_001168247.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100203557.hg.1 | NM_001300859.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100203557.hg.1 | NM_144990.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100203557.hg.1 | TC0100013853.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000302946.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000302946.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000302946.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000302946.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000302946.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000302946.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000302946.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000302946.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000359345.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000372611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000372611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000372611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000372611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000372611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000372611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000372611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000372611.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000372613.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000372613.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000372613.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000372613.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000372613.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000372613.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000372613.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000372613.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000372613.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000372613.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000372613.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000439569.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000439569.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000439569.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000439569.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000439569.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000439569.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000439569.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000439569.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000439569.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | ENST00000439569.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_001168247.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_001168247.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_001168247.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_001168247.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_001168247.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_001168247.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_001168247.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_001168247.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_001168247.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_001168247.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_001300859.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_001300859.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_001300859.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_001300859.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_001300859.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_001300859.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_001300859.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_001300859.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_144990.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_144990.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_144990.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_144990.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_144990.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_144990.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_144990.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013853.hg.1 | NM_144990.3 | Refseq |

SLFNL1
Genomic Map