Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | HOXC cluster antisense RNA 3 | HOXC-AS3 | -326.0 | Anti-sense | Chr. 12 - 53983951 - 53985519 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC440101 | FLJ12825 | 23.4kb | Sense | Chr. 12 - 54058254 - 54122234 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map