Assay Details
| Gene Symbol: USP17L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201402.2 | NP_958804.2 | 1 | 508 | 157 | |
| GenBank mRNA | AY509884.1 | - | 1 | 508 | 157 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cell cycle checkpoint
ubiquitin-dependent protein catabolic process
apoptotic process
mitotic cell cycle checkpoint
negative regulation of protein processing
protein deubiquitination
regulation of cell migration
negative regulation of histone deacetylation
negative regulation of GTPase activity
regulation of cell proliferation
regulation of apoptotic process
positive regulation of GTPase activity
regulation of defense response to virus by host
protein K63-linked deubiquitination
protein K48-linked deubiquitination
CAAX-box protein processing
negative regulation of protein targeting to membrane
regulation of ruffle assembly
positive regulation of MDA-5 signaling pathway
positive regulation of RIG-I signaling pathway
thiol-dependent ubiquitin-specific protease activity
protein binding
cell cycle checkpoint
ubiquitin-dependent protein catabolic process
apoptotic process
mitotic cell cycle checkpoint
negative regulation of protein processing
protein deubiquitination
regulation of cell migration
negative regulation of histone deacetylation
negative regulation of GTPase activity
regulation of cell proliferation
regulation of apoptotic process
positive regulation of GTPase activity
regulation of defense response to virus by host
protein K63-linked deubiquitination
protein K48-linked deubiquitination
CAAX-box protein processing
negative regulation of protein targeting to membrane
regulation of ruffle assembly
positive regulation of MDA-5 signaling pathway
positive regulation of RIG-I signaling pathway
thiol-dependent ubiquitin-specific protease activity
protein binding
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26239778 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26239727 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16069649 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26239805 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3292739 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3342153 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3260451 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3449 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P272917 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3450 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 52890 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 3635 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 4093 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3342153 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3260451 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3292739 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3260451 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3292739 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3342153 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16069739 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26239778 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26239805 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26239787 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26239772 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26239778 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16069739 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26239772 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26239787 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26239805 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P272917 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P05959799 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8149244 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8094117 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8149357 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8094121 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8094119 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8144398 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8149242 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8094131 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8094129 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8094133 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8094123 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8094127 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8144396 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8094125 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8094130 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8094126 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8094128 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8094118 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8094120 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8094116 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8144395 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8094124 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8094132 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8149241 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8149243 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8149356 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8144397 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009493.hg.1 | ENST00000527080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009493.hg.1 | ENST00000527080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009493.hg.1 | ENST00000530484.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009493.hg.1 | ENST00000530484.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009493.hg.1 | ENST00000625190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009493.hg.1 | ENST00000625190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009493.hg.1 | NM_001256871.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009493.hg.1 | NM_001256871.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009493.hg.1 | NM_001256872.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009493.hg.1 | NM_001256872.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009651.hg.1 | ENST00000526502.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009651.hg.1 | ENST00000526502.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009651.hg.1 | ENST00000526502.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009651.hg.1 | NR_046415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009651.hg.1 | ENST00000526502.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009651.hg.1 | ENST00000526502.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009651.hg.1 | ENST00000526502.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009651.hg.1 | NR_046415.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009493.hg.1 | ENST00000527080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009493.hg.1 | ENST00000527080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009493.hg.1 | ENST00000530484.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009493.hg.1 | ENST00000530484.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009493.hg.1 | ENST00000625190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009493.hg.1 | ENST00000625190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009493.hg.1 | NM_001256871.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009493.hg.1 | NM_001256871.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009493.hg.1 | NM_001256872.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009493.hg.1 | NM_001256872.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035531 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035531 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_2335123 | NM_001242332.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2335123 | NM_001242331.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2335123 | NM_001242330.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2335123 | NM_001242328.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2335123 | NM_001242329.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1726758 | NM_001242332.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1726758 | NM_001242331.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1726758 | NM_001242330.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1726758 | NM_001242328.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1726758 | NM_001242329.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2335123 | NM_001242332.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2335123 | NM_001242331.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2335123 | NM_001242330.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2335123 | NM_001242328.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2335123 | NM_001242329.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2335123 | NM_001242332.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2335123 | NM_001242331.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2335123 | NM_001242330.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2335123 | NM_001242328.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2335123 | NM_001242329.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2335123 | NM_001242332.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2335123 | NM_001242331.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2335123 | NM_001242330.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2335123 | NM_001242328.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2335123 | NM_001242329.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_41349931-S | BC100992.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_41349931-S | BC100993.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_41349931-S | AF544012.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_41349931-S | AY188990.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_41349931-S | BC100991.2 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_5668-A | - | - |
| Illumina human-6 v1.0 expression beadchip | 0006290739 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0006290739 | - | - |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 3520753 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_5668 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2335123 | NM_001242332.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2335123 | NM_001242331.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2335123 | NM_001242330.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2335123 | NM_001242328.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2335123 | NM_001242329.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2335123 | NM_001242332.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2335123 | NM_001242331.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2335123 | NM_001242330.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2335123 | NM_001242328.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2335123 | NM_001242329.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1726758 | NM_001242332.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1726758 | NM_001242331.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1726758 | NM_001242330.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1726758 | NM_001242328.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1726758 | NM_001242329.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2335123 | NM_001242332.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2335123 | NM_001242331.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2335123 | NM_001242330.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2335123 | NM_001242328.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2335123 | NM_001242329.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2335123 | NM_001242332.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2335123 | NM_001242331.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2335123 | NM_001242330.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2335123 | NM_001242328.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2335123 | NM_001242329.1 | human_transcriptome |

USP17L2
Genomic Map