Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100506271 | LOC100506271 | -16.8kb | Anti-sense | Chr. 22 - 37352190 - 37354839 on NCBI Build GRCh38 | |
| Downstream | MFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase | MFNG | 96.4kb | Anti-sense | Chr. 22 - 37469063 - 37486440 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map