Assay Details
| Gene Symbol: PA2G4P4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003284.1 | 1 | 2459 | 158 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02699740 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22839877 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22839864 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22839879 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22839846 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22839876 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22839878 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22839851 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02699743 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02699751 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22839845 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22839869 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02699756 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22839852 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22839854 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02699742 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22839848 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22839872 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22839853 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22839861 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22839866 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22839870 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02699737 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02699754 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02699766 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22839856 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02699760 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22839867 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02699752 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22839855 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22839874 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02699753 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 69312 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P827491 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3227209 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699777 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699795 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839905 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839856 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699793 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839906 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699754 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699751 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839898 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839870 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699752 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839843 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839896 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839854 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699756 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699763 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839874 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839904 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699783 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699733 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839866 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839877 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15503319 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839851 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839892 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839876 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699778 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699786 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839867 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699734 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699736 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839897 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839873 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839855 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839845 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699742 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839878 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839861 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699753 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839887 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839872 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839852 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699770 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699757 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839872 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699757 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839843 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839896 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699786 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699778 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699763 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839898 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839906 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839892 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839876 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699777 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839877 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839854 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839867 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699756 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699734 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839855 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839897 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699751 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699736 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839905 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839887 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699795 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699793 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839874 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699753 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699752 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839851 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699742 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839873 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699783 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839845 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839866 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839878 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699770 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839856 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839852 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699754 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02699733 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839904 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839870 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15503319 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22839861 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 4918 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P473834 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699789 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699697 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699780 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699813 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15503309 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699743 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699776 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699817 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699703 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699787 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699714 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699712 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699771 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699704 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699734 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699737 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699725 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699675 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699755 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699808 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15503308 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699677 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699679 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699717 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699747 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699806 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699764 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699758 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15503324 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699774 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699821 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699753 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699799 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699730 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699802 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699766 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699793 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699811 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699722 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699727 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15503331 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699782 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15503307 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699710 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699708 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699795 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699815 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699683 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699691 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699762 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699791 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15503328 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699689 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699720 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699750 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699700 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699686 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699784 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699761 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699739 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699694 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699777 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699797 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699768 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02699706 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Clariom D Assay | PSR0300196461.hg.1 | NR_003284.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196461.hg.1 | NR_003284.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196461.hg.1 | TC0300012908.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 216422_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216422_PM_at | - | - |
| Human Genome U133A 2.0 Array | 216422_PM_at | - | - |
| Human Genome U133A 2.0 Array | 216422_at | - | - |
| Human Genome U133 Set | 216422_at | - | - |
| Human Genome U133 Set | 216422_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A021835 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A021835 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_2181748 | NR_003284.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2181748 | NR_003284.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2181748 | NR_003284.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2181748 | NR_003284.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2181748 | NR_003284.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2181748 | NR_003284.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2181748 | NR_003284.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2181748 | NR_003284.1 | human_transcriptome |

PA2G4P4
Genomic Map