Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small nucleolar RNA, C/D box 89 |
SNORD89 | HBII-289 | -1.2kb | Sense | Chr. 2 - 101272936 - 101273049 on NCBI Build GRCh38 |
| Downstream | microRNA 5696 | MIR5696 | 34.6kb | Anti-sense | Chr. 2 - 101309450 - 101309534 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map