Assay Details
| Gene Symbol: UQCRH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297565.1 | NP_001284494.1 | 1 | 60 | 67 | |
| NM_001297566.1 | NP_001284495.1 | 1 | 60 | 67 | ||
| NM_006004.3 | NP_005995.2 | 1 | 60 | 67 | ||
| GenBank mRNA | BP366403.1 | - | 1 | 62 | 67 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000583.hg
Additional Information:
This assay is in the 5'UTR region of the NM_001297565.1,NM_001297566.1,NM_006004.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100013013.hg.1 | ENST00000483273.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013013.hg.1 | ENST00000483273.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013013.hg.1 | ENST00000483273.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013013.hg.1 | ENST00000483273.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013013.hg.1 | NM_001089591.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013013.hg.1 | NM_001089591.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013013.hg.1 | NM_001089591.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01009937.hg.1 | ENST00000311672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01009937.hg.1 | ENST00000486951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01009937.hg.1 | NM_006004.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01009937.hg.1 | TC01000583.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01009938.hg.1 | ENST00000311672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01009938.hg.1 | ENST00000486951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01009938.hg.1 | NM_006004.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01009938.hg.1 | TC01000583.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01009944.hg.1 | ENST00000311672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01009944.hg.1 | NM_006004.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01009944.hg.1 | TC01000583.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000311672.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000311672.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000311672.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000311672.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000311672.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000311672.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000311672.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000460947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000460947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000460947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000460947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000460947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000486951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000486951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000486951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000486951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000489056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000489056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000489056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000489056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000489056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000489056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000496387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000496387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000496387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000496387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000496387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000496387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | ENST00000496387.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | NM_001297565.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | NM_001297565.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | NM_001297565.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | NM_001297565.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | NM_001297565.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | NM_001297565.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | NM_001297565.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | NM_001297565.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | NM_001297566.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | NM_001297566.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | NM_001297566.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | NM_001297566.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | NM_001297566.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | NM_001297566.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | NM_006004.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | NM_006004.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | NM_006004.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | NM_006004.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | NM_006004.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | NM_006004.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008152.hg.1 | NM_006004.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013013.hg.1 | ENST00000483273.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013013.hg.1 | ENST00000483273.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013013.hg.1 | ENST00000483273.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013013.hg.1 | ENST00000483273.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013013.hg.1 | NM_001089591.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013013.hg.1 | NM_001089591.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013013.hg.1 | NM_001089591.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158320.hg.1 | ENST00000311672.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158320.hg.1 | ENST00000486951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158320.hg.1 | ENST00000489056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158320.hg.1 | ENST00000496387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158320.hg.1 | NM_001297565.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158320.hg.1 | NM_001297566.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158320.hg.1 | NM_006004.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158320.hg.1 | ENST00000311672.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158320.hg.1 | ENST00000486951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158320.hg.1 | ENST00000489056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158320.hg.1 | ENST00000496387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158320.hg.1 | NM_001297565.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158320.hg.1 | NM_001297566.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158320.hg.1 | NM_006004.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158320.hg.1 | TC0100008152.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000311672.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000311672.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000311672.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000311672.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000311672.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000311672.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000311672.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000460947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000460947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000460947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000460947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000460947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000486951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000486951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000486951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000486951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000489056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000489056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000489056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000489056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000489056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000489056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000496387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000496387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000496387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000496387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000496387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000496387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | ENST00000496387.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | NM_001297565.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | NM_001297565.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | NM_001297565.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | NM_001297565.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | NM_001297565.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | NM_001297565.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | NM_001297565.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | NM_001297565.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | NM_001297566.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | NM_001297566.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | NM_001297566.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | NM_001297566.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | NM_001297566.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | NM_001297566.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | NM_006004.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | NM_006004.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | NM_006004.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | NM_006004.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | NM_006004.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | NM_006004.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008152.hg.1 | NM_006004.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158313.hg.1 | ENST00000311672.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158313.hg.1 | NM_001297565.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158313.hg.1 | NM_001297566.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158313.hg.1 | NM_006004.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158313.hg.1 | ENST00000311672.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158313.hg.1 | NM_001297565.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158313.hg.1 | NM_001297566.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158313.hg.1 | NM_006004.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158313.hg.1 | TC0100008152.hg.1 | Clariom D TC |

UQCRH
Genomic Map