Assay Details
| Gene Symbol: RPS10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203245.2 | NP_001190174.1 | 1 | 51 | 149 | |
| GenBank mRNA | BG532491.1 | - | 1 | 52 | 149 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06004130.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001203245.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ribosomal small subunit assembly
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
structural constituent of ribosome
protein binding
poly(A) RNA binding
ribosomal small subunit assembly
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
structural constituent of ribosome
protein binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06019580.hg.1 | ENST00000494077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06019580.hg.1 | NM_001203245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06019580.hg.1 | TC06004130.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000326199.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000326199.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000326199.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000326199.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000326199.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000326199.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000326199.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000326199.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000344700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000344700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000344700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000344700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000344700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000344700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000344700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000464218.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000464218.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000464218.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000464218.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000464218.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000464218.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000464218.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000467531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000467531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000467531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000467531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000467531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000467531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000467531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000480942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000480942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000480942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000494077.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000494077.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000494077.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000494077.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000494077.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000494077.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000494077.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000621356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000621356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000621356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000621356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000621356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000621356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | ENST00000621356.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | NM_001014.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | NM_001014.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | NM_001014.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | NM_001014.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | NM_001014.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | NM_001014.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | NM_001014.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | NM_001014.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | NM_001203245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | NM_001203245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | NM_001203245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | NM_001203245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | NM_001203245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | NM_001203245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | NM_001203245.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | NM_001204091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | NM_001204091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | NM_001204091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | NM_001204091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | NM_001204091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | NM_001204091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | NM_001204091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014282.hg.1 | NM_001204091.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008203.hg.1 | ENST00000480396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008203.hg.1 | ENST00000480396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000326199.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000326199.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000326199.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000326199.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000326199.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000326199.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000326199.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000326199.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000344700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000344700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000344700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000344700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000344700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000344700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000344700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000464218.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000464218.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000464218.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000464218.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000464218.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000464218.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000464218.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000467531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000467531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000467531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000467531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000467531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000467531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000467531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000480942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000480942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000480942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000494077.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000494077.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000494077.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000494077.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000494077.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000494077.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000494077.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000621356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000621356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000621356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000621356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000621356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000621356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | ENST00000621356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | NM_001014.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | NM_001014.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | NM_001014.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | NM_001014.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | NM_001014.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | NM_001014.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | NM_001014.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | NM_001014.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | NM_001203245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | NM_001203245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | NM_001203245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | NM_001203245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | NM_001203245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | NM_001203245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | NM_001203245.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | NM_001204091.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | NM_001204091.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | NM_001204091.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | NM_001204091.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | NM_001204091.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | NM_001204091.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | NM_001204091.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014282.hg.1 | NM_001204091.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201630.hg.1 | ENST00000326199.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201630.hg.1 | ENST00000344700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201630.hg.1 | ENST00000464218.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201630.hg.1 | ENST00000467531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201630.hg.1 | ENST00000480942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201630.hg.1 | ENST00000494077.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201630.hg.1 | ENST00000621356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201630.hg.1 | NM_001014.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201630.hg.1 | NM_001203245.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201630.hg.1 | NM_001204091.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201630.hg.1 | ENST00000326199.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201630.hg.1 | ENST00000344700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201630.hg.1 | ENST00000464218.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201630.hg.1 | ENST00000467531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201630.hg.1 | ENST00000480942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201630.hg.1 | ENST00000494077.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201630.hg.1 | ENST00000621356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201630.hg.1 | NM_001014.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201630.hg.1 | NM_001203245.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201630.hg.1 | NM_001204091.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201630.hg.1 | TC0600014282.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600201634.hg.1 | ENST00000494077.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201634.hg.1 | ENST00000621356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201634.hg.1 | NM_001203245.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201634.hg.1 | ENST00000494077.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201634.hg.1 | ENST00000621356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201634.hg.1 | NM_001203245.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201634.hg.1 | TC0600014282.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600201629.hg.1 | ENST00000326199.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201629.hg.1 | ENST00000344700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201629.hg.1 | ENST00000464218.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201629.hg.1 | ENST00000467531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201629.hg.1 | ENST00000480942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201629.hg.1 | ENST00000494077.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201629.hg.1 | ENST00000621356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201629.hg.1 | NM_001014.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201629.hg.1 | NM_001203245.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201629.hg.1 | NM_001204091.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201629.hg.1 | ENST00000326199.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201629.hg.1 | ENST00000344700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201629.hg.1 | ENST00000464218.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201629.hg.1 | ENST00000467531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201629.hg.1 | ENST00000480942.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201629.hg.1 | ENST00000494077.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201629.hg.1 | ENST00000621356.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201629.hg.1 | NM_001014.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201629.hg.1 | NM_001203245.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201629.hg.1 | NM_001204091.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201629.hg.1 | TC0600014282.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1900008203.hg.1 | ENST00000480396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008203.hg.1 | ENST00000480396.1 | Ensembl |

RPS10
Genomic Map