Assay Details
| Gene Symbol: RNASE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002933.4 | NP_002924.1 | 2 | 773 | 135 | |
| NM_198232.2 | NP_937875.1 | 2 | 843 | 135 | ||
| NM_198234.2 | NP_937877.1 | 3 | 844 | 135 | ||
| NM_198235.2 | NP_937878.1 | 3 | 856 | 135 | ||
| GenBank mRNA | AI400223.1 | - | 1 | 135 | ||
| AK098375.1 | - | 2 | 823 | 135 | ||
| AK129871.1 | - | 2 | 764 | 135 | ||
| BC005324.1 | - | 2 | 773 | 3950230 | 135 | |
| BC022882.1 | - | 2 | 747 | 4294153 | 135 | |
| BM919747.1 | - | 2 | 728 | 135 | ||
| BP385178.1 | - | 1 | 535 | 135 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RNASE1
Genomic Map