Assay Details
| Gene Symbol: DSCR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AA770014.1 | - | 1 | 47 | 121 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000357704.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000357704.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000357704.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000357704.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000400477.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000400477.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000400477.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000400477.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000465532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000465532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000465532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000465532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000465532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000465532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000469658.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000469658.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000469658.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000478613.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000478613.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000478613.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000478613.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000478613.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000495344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000495344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000495344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000495344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000495344.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000614538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000614538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | ENST00000614538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | NR_026838.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | NR_026838.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | NR_026838.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | NR_026838.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | NR_026838.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | NR_026838.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | NR_026839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | NR_026839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | NR_026839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | NR_026839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | NR_026839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | NR_026840.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | NR_026840.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | NR_026840.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | NR_026840.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | NR_026841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | NR_026841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | NR_026841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | NR_026841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | NR_026842.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | NR_026842.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008509.hg.1 | NR_026842.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000357704.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000357704.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000357704.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000357704.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000400477.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000400477.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000400477.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000400477.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000465532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000465532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000465532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000465532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000465532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000465532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000469658.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000469658.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000469658.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000478613.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000478613.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000478613.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000478613.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000478613.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000495344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000495344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000495344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000495344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000495344.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000614538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000614538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | ENST00000614538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | NR_026838.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | NR_026838.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | NR_026838.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | NR_026838.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | NR_026838.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | NR_026838.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | NR_026839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | NR_026839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | NR_026839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | NR_026839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | NR_026839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | NR_026840.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | NR_026840.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | NR_026840.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | NR_026840.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | NR_026841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | NR_026841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | NR_026841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | NR_026841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | NR_026842.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | NR_026842.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008509.hg.1 | NR_026842.1 | Refseq |

DSCR8
Genomic Map